Siramesine hydrochloride ≥98.0%

Leverantör: TCI

I1090-25MG I1090-5MG
TCIAI1090-25MGEA 2125 SEK
TCIAI1090-25MG TCIAI1090-5MG
Siramesine hydrochloride ≥98.0%
Siramesine hydrochloride
Formel: C₃₀H₃₁FN₂O·HCl
Storage Temperature: Frys
MDL Number: MFCD28144522
CAS nummer: 224177-60-0

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 223.0 to 227.0 deg-C
Elemental analysis (Nitrogen) gt. 5.20 to 5.80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR