2-Metylpropen ∼9% i hexan

Leverantör: TCI EUROPE
Fara

Synonyms: 2-Methylpropene, Isobutene, Isobutylene

I1083-100ML I1083-500ML
TCIAI1083-100MLEA 335 SEK
TCIAI1083-100ML TCIAI1083-500ML
2-Metylpropen ∼9% i hexan
2-Metylpropen
Formel: C₄H₈
MW: 56,11 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00008898
CAS nummer: 115-11-7
EINECS: 204-066-3
UN: 1993
ADR: 3,II

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 9.0 to 11.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR