2-Isopropyl-4-(N-methylaminomethyl)thiazole dihydrochloride ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
1-(2-Isopropylthiazol-4-yl)-N-methylmethanamine dihydrochloride
I1057-1G
I1057-5G
TCIAI1057-1GEA
525
SEK
TCIAI1057-1G
TCIAI1057-5G
2-Isopropyl-4-(N-methylaminomethyl)thiazole dihydrochloride ≥98.0% (by HPLC, titration analysis)
2-Isopropyl-4-(N-methylaminomethyl)thiazole dihydrochloride
Formel:
C₈H₁₄N₂S·₂HCl MW: 243,19 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD11841218 CAS nummer: 1185167-55-8 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...