3-Iodobiphenyl ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
3-Iodo-biphenyl
I1055-1G
I1055-5G
TCIAI1055-1GEA
750
SEK
TCIAI1055-1G
TCIAI1055-5G
3-Iodobiphenyl ≥98.0%
3-Iodobiphenyl
Formel:
C₁₂H₉I MW: 280,11 g/mol Kokpunkt: 178°C |
MDL Number:
MFCD09032441 CAS nummer: 20442-79-9 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...