4-Iodo-3-nitrotoluene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1-Iodo-4-methyl-2-nitrobenzene

I1022-1G I1022-5G
TCIAI1022-1GEA 270 SEK
TCIAI1022-1G TCIAI1022-5G
4-Iodo-3-nitrotoluene ≥98.0%
4-Iodo-3-nitrotoluene
Formel: C₇H₆INO₂
MW: 263,03 g/mol
MDL Number: MFCD00024196
CAS nummer: 5326-39-6

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR