3-Iodo-6-nitro-1H-indazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-Iodo-6-nitroindazole
I1004-1G
I1004-5G
TCIAI1004-1GEA
1125
SEK
TCIAI1004-1G
TCIAI1004-5G
3-Iodo-6-nitro-1H-indazole ≥98.0%
3-Iodo-6-nitro-1H-indazole
Formel:
C₇H₄IN₃O₂ MW: 289,0300 g/mol |
MDL Number:
MFCD07781652 CAS nummer: 70315-70-7 |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...