4H-Indeno[1,2-b]thiophen-4-one ≥98.0%
Leverantör: TCI EUROPE
I1003-200MG
I1003-1G
TCIAI1003-200MGEA
575
SEK
TCIAI1003-200MG
TCIAI1003-1G
4H-Indeno[1,2-b]thiophen-4-one ≥98.0%
4H-Indeno[1,2-b]thiophen-4-one
Formel:
C₁₁H₆OS MW: 186,23 g/mol |
CAS nummer:
5706-08-1 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 99.0 to 103.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...