Isopropylhydrazine hydrochloride ≥98.0%

Leverantör: TCI
Varning

I0964-1G I0964-5G
TCIAI0964-1GEA 390 SEK
TCIAI0964-1G TCIAI0964-5G
Isopropylhydrazine hydrochloride ≥98.0%
Isopropylhydrazine hydrochloride
Formel: C₃H₁₁ClN₂
MDL Number: MFCD01746887
CAS nummer: 16726-41-3
EINECS: 240-784-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Potassium iodate Method) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR