Copper(II)-2-ethylhexanoate
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Ethylhexanoic acid copper (II) salt, Copper(II) 2-ethylhexanoate, 2-Ethylcaproic acid copper (II) salt, Copper bis(2-ethylhexanoate)
20539.09 20539.18
20539.09EA 1725 SEK
20539.09 20539.18
Copper(II)-2-ethylhexanoate
Copper(II)-2-ethylhexanoate
CAS No.: 149-11-1 Molecular Formula: C16H30CuO4 Formula Weight: 349.96 Physical Form: Powder/Lump Appearance: Blue-green Melting Point: 252° dec. MDL No.: MFCD00015695
Formel: C₁₆H₃₀CuO₄
MW: 349,96 g/mol
Smältpunkt: 252 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00015695
CAS nummer: 149-11-1
EINECS: 205-731-0