5-Iodo-2-methoxypyridine ≥98.0%
Leverantör: TCI EUROPE
Varning
I0925-1G I0925-5G
TCIAI0925-1GEA 550 SEK
TCIAI0925-1G TCIAI0925-5G
5-Iodo-2-methoxypyridine ≥98.0%
5-Iodo-2-methoxypyridine
Formel: C₆H₆INO
MW: 235,03 g/mol
Kokpunkt: 74°C
MDL Number: MFCD07781180
CAS nummer: 13472-61-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c893c557d79a95fa9b36ba75c79eccc54c013c9d&vid=c893c557d79a95fa9b36ba75c79eccc54c013c9d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %