3-Iodocarbazole ≥98.0%

Leverantör: TCI
Varning

Synonyms: 3-Iodo-9H-carbazole

I0919-5G I0919-25G
TCIAI0919-5GEA 550 SEK
TCIAI0919-5G TCIAI0919-25G
3-Iodocarbazole ≥98.0%
3-Iodocarbazole
Formel: C₁₂H₈IN
MW: 293.11 g/mol
MDL Number: MFCD00267746
CAS nummer: 16807-13-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86787ecc2325ab428f7e88c5fe7eee8df2f6d9f2&vid=86787ecc2325ab428f7e88c5fe7eee8df2f6d9f2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 197.0 to 201.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR