5-Iodo-o-toluic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
5-lodo-2-methylbenzoic acid, 5-Iodo-2-methylbenzoic acid
I0916-5G
I0916-25G
TCIAI0916-5GEA
245
SEK
TCIAI0916-5G
TCIAI0916-25G
5-Iodo-o-toluic acid ≥98.0% (by GC, titration analysis)
5-Iodo-o-toluic acid
Formel:
C₈H₇IO₂ MW: 262,0400 g/mol |
MDL Number:
MFCD01570258 CAS nummer: 54811-38-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 179.0 to 183.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...