3-Iodo-9-phenylcarbazole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Iodo-N-phenylcarbazole

I0913-1G I0913-5G
TCIAI0913-1GEA 215 SEK
TCIAI0913-1G TCIAI0913-5G
3-Iodo-9-phenylcarbazole ≥98.0%
3-Iodo-9-phenylcarbazole
Formel: C₁₈H₁₂IN
MW: 369,21 g/mol
MDL Number: MFCD09261276
CAS nummer: 502161-03-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43a8aeddde3209952735882cf62482e39bbdc764&vid=43a8aeddde3209952735882cf62482e39bbdc764&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 103.0 to 107.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR