Ipratropium bromide monohydrate ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

Synonyms: Atropine Isopropyl Bromide

I0907-200MG I0907-1G
TCIAI0907-200MGEA 1050 SEK
TCIAI0907-200MG TCIAI0907-1G
Ipratropium bromide monohydrate ≥98.0% (by HPLC, total nitrogen)
Ipratropium bromide monohydrate
Formel: C₂₀H₃₀BrNO₃
MW: 412,37 g/mol
MDL Number: MFCD09842549
CAS nummer: 66985-17-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Water 3.5 to 4.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR