2-Iodo-9,9-dimethylfluorene ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2-Iodo-9,9-dimethyl-9H-fluorene

I0875-5G I0875-25G
TCIAI0875-5GEA 1425 SEK
TCIAI0875-5G TCIAI0875-25G
2-Iodo-9,9-dimethylfluorene ≥98.0%
2-Iodo-9,9-dimethylfluorene
Formel: C₁₅H₁₃I
MW: 320,17 g/mol
MDL Number: MFCD12024272
CAS nummer: 144981-85-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f9031c35f1bf61bbe9dfca282b403a52314da87&vid=9f9031c35f1bf61bbe9dfca282b403a52314da87&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR