Indole-2-carbaldehyd ≥97.0%

Leverantör: TCI
Varning

Synonyms: Indole-2-carboxaldehyde, 1H-Indole-2-carbaldehyde

I0852-1G I0852-5G
TCIAI0852-1GEA 845 SEK
TCIAI0852-1G TCIAI0852-5G
Indole-2-carbaldehyd ≥97.0%
Indole-2-carbaldehyd
Formel: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD03001425
CAS nummer: 19005-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36525e59a72ff7c92595e91253b82000f853fc16&vid=36525e59a72ff7c92595e91253b82000f853fc16&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 138.0 to 142.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR