L-Isoserine ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
(S)-3-Amino-2-hydroxypropanoic acid, L-Isoserine, (S)-3-Amino-2-hydroxypropionic Acid
I0828-1G
I0828-5G
TCIAI0828-1GEA
230
SEK
TCIAI0828-1G
TCIAI0828-5G
L-Isoserine ≥98.0%
L-Isoserine
Formel:
C₃H₇NO₃ MW: 105,09 g/mol |
MDL Number:
MFCD03428326 CAS nummer: 632-13-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -31.0 to -34.0 deg (C=1, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...