Imidazo[1,2-a]pyrimidine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
I0798-1G
I0798-5G
TCIAI0798-1GEA
1450
SEK
TCIAI0798-1G
TCIAI0798-5G
Imidazo[1,2-a]pyrimidine ≥98.0%
Imidazo[1,2-a]pyrimidine
Formel:
C₆H₅N₃ MW: 119,1200 g/mol |
MDL Number:
MFCD06245377 CAS nummer: 274-95-3 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Melting point | 125.0 to 129.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...