2-Iodoresorcinol ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,3-Dihydroxy-2-iodobenzene, 2-Iodobenzene-1,3-diol

I0788-1G I0788-5G
TCIAI0788-1GEA 1225 SEK
TCIAI0788-1G TCIAI0788-5G
2-Iodoresorcinol ≥97.0%
2-Iodoresorcinol
Formel: C₆H₅IO₂
MW: 236,01 g/mol
Storage Temperature: Frys
MDL Number: MFCD09701440
CAS nummer: 41046-67-7

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 106.0 to 110.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR