3,5-Bis(trifluoromethyl)phenyl Isocyanate ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 1-Isocyanato-3,5-bis(trifluoromethyl)benzene, 3,5-Bis(trifluoromethyl)phenylisocyanate, Isocyanic Acid 3,5-Bis(trifluoromethyl)phenyl Ester
I0766-25G I0766-5G
TCIAI0766-25GEA 4450 SEK
TCIAI0766-25G TCIAI0766-5G
3,5-Bis(trifluoromethyl)phenyl Isocyanate ≥98.0%
3,5-Bis(trifluoromethyl)phenyl Isocyanate
Formel: C₉H₃F₆NO
MW: 255,12 g/mol
Kokpunkt: 78°C
Flampunkt: 68°C
MDL Number: MFCD00013559
CAS nummer: 16588-74-2
UN: 2206
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %