4-(Isopropylamino)butanol ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
I0756-1G
I0756-5G
TCIAI0756-1GEA
415
SEK
TCIAI0756-1G
TCIAI0756-5G
4-(Isopropylamino)butanol ≥98.0% (by GC, titration analysis)
4-(Isopropylamino)butanol
Formel:
C₇H₁₇NO MW: 131,22 g/mol Kokpunkt: 85°C |
MDL Number:
MFCD14708173 CAS nummer: 42042-71-7 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...