Ibudilast ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI
Varning
I0740-100MG I0740-20MG
TCIAI0740-100MGEA 590 SEK
TCIAI0740-100MG TCIAI0740-20MG
Ibudilast ≥98.0% (by HPLC, titration analysis)
Ibudilast
Formel: C₁₄H₁₈N₂O
Kokpunkt: 175°C
Storage Temperature: Kylskåp
MDL Number: MFCD00864808
CAS nummer: 50847-11-5
Merck Index: 14,04879
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 54.0 to 58.0 deg-C