4-Isopropenyltoluene ≥95.0% stabilised
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4,alpha-Dimethylstyrene, 1-Isopropenyl-4-methylbenzene
Stabilization:
med 4-tert-Butylpyrokatekol
I0693-25G
I0693-5G
TCIAI0693-25GEA
5225
SEK
TCIAI0693-25G
TCIAI0693-5G
4-Isopropenyltoluene ≥95.0% stabilised
4-Isopropenyltoluene
Formel:
C₁₀H₁₂ MW: 132,21 g/mol Kokpunkt: 186°C Smältpunkt: –28°C Flampunkt: 62°C Storage Temperature: Frys |
MDL Number:
MFCD00036510 CAS nummer: 1195-32-0 EINECS: 214-795-9 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...