3-Iodo-2-propynyl-N-butylcarbamate ≥97.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3-Iodoprop-2-yn-1-yl butylcarbamate, N-Butylcarbamic Acid 3-Iodo-2-propynyl Ester, IPBC
I0666-25G
I0666-100G
TCIAI0666-25GEA
565
SEK
TCIAI0666-25G
TCIAI0666-100G
3-Iodo-2-propynyl-N-butylcarbamate ≥97.0%
3-Iodo-2-propynyl-N-butylcarbamate
Formel:
C₈H₁₂INO₂ MW: 281,09 g/mol |
MDL Number:
MFCD00072438 CAS nummer: 55406-53-6 EINECS: 259-627-5 UN: 2811 ADR: 6.1,III Merck Index: 14,05069 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 67.0 to 71.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...