Isobornyl acrylate ≥90.0% stabilised
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(1R,2R,4R)-rel-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl acrylate, Acrylic acid Isobornyl Ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
I0638-25G
I0638-500G
TCIAI0638-25GEA
230
SEK
TCIAI0638-25G
TCIAI0638-500G
Isobornyl acrylate ≥90.0% stabilised
Isobornyl acrylate
Formel:
C₁₃H₂₀O₂ MW: 208,30 g/mol Kokpunkt: 104°C Flampunkt: 109°C |
MDL Number:
MFCD00080424 CAS nummer: 5888-33-5 EINECS: 227-561-6 UN: 3082 ADR: 9,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...