2-Iodobenzaldehyde ≥98.0%

Leverantör: TCI EUROPE
Varning

I0610-1G I0610-5G
TCIAI0610-1GEA 845 SEK
TCIAI0610-1G TCIAI0610-5G
2-Iodobenzaldehyde ≥98.0%
2-Iodobenzaldehyde
Formel: C₇H₅IO
MW: 232,02 g/mol
Kokpunkt: 102°C
Storage Temperature: Kylskåp
MDL Number: MFCD00039570
CAS nummer: 26260-02-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14606053b1bec1bf69370736f273c6245b2fd47f&vid=14606053b1bec1bf69370736f273c6245b2fd47f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR