(S)-(+)-2-(4-Isobutylphenyl)propionic Acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: (S)-2-(4-Isobutylphenyl)propanoic acid, (S)-(+)-4-Isobutyl-alpha-methylphenylacetic Acid, (S)-(+)-Ibuprofen

I0549-1G I0549-5G
TCIAI0549-1GEA 430 SEK
TCIAI0549-1G TCIAI0549-5G
(S)-(+)-2-(4-Isobutylphenyl)propionic Acid ≥98.0% (by GC, titration analysis)
(S)-(+)-2-(4-Isobutylphenyl)propionic Acid
Formel: C₁₃H₁₈O₂
MW: 206,29 g/mol
MDL Number: MFCD00069289
CAS nummer: 51146-56-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 51.0 to 55.0 deg-C
Specific rotation [a]20/D +58.0 to +60.0 deg (C=2, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR