3-Isochromanone ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,4-Dihydro-3H-2-benzopyran-3-one

I0544-1G I0544-25G I0544-5G
TCIAI0544-1GEA 325 SEK
TCIAI0544-1G TCIAI0544-25G TCIAI0544-5G
3-Isochromanone ≥99.0%
3-Isochromanone
Formel: C₉H₈O₂
MW: 148,16 g/mol
Kokpunkt: 130°C
MDL Number: MFCD00043005
CAS nummer: 4385-35-7
EINECS: 224-493-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystaline
Purity (GC) min. 99.0 %
Melting point 82.0 to 84.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR