Indocyanine green

Leverantör: TCI EUROPE
Varning

Synonyms: IR-125, 4,5-benzoindotricarbocyanine, IC Green, Cardiogreen, Sodium 4-(2-(7-(1,1-dimethyl-3-(4-sulphonatobutyl)-1H-benzo[e]indol-2(3H)-ylidene)hepta-1,3,5-trien-1-yl)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulphonate, Foxgreen

I0535-1G I0535-100MG
TCIAI0535-1GEA 7175 SEK
TCIAI0535-1G TCIAI0535-100MG
Indocyanine green
Indocyanine green
Formel: C₄₃H₄₇N₂NaO₆S₂
MW: 774,99 g/mol
MDL Number: MFCD00013078
CAS nummer: 3599-32-4
EINECS: 222-751-5
Merck Index: 14,04962

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green powder to crystal
Lambda max. 785.0 to 789.0 nm (EtOH)
Absorbance (E1%1cm) min. 2000 (EtOH, 785.0 to 789.0 nm)
Drying loss max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR