1-Iodo-2-methylbutane ≥95.0% stabilised
Leverantör: TCI EUROPE
Varning
Stabilization: Stabilised with copper chip
I0530-25G I0530-5G
TCIAI0530-25GEA 6050 SEK
TCIAI0530-25G TCIAI0530-5G
1-Iodo-2-methylbutane ≥95.0% stabilised
1-Iodo-2-methylbutane
Formel: C₅H₁₁I
MW: 198,05 g/mol
Kokpunkt: 148°C
Flampunkt: 42°C
MDL Number: MFCD01321177
CAS nummer: 616-14-8
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Colorless to Brown clear liquid
Purity (GC) min. 95.0 %