Isovaleraldehyde 2,4-Dinitrophenylhydrazone ≥99.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
I0477-1G
I0477-10G
TCIAI0477-1GEA
430
SEK
TCIAI0477-1G
TCIAI0477-10G
Isovaleraldehyde 2,4-Dinitrophenylhydrazone ≥99.0% (by HPLC, titration analysis)
Isovaleraldehyde 2,4-Dinitrophenylhydrazone
Formel:
C₁₁H₁₄N₄O₄ MW: 266,26 g/mol |
MDL Number:
MFCD00209593 CAS nummer: 2256-01-1 UN: 1325 ADR: 4.1,III |
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 126.0 to 129.0 deg-C |
Solubility in Ethylacetate | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...