Methyl (S)-(-)-2-isocyanato-4-methylvalerate ≥98.0%

Leverantör: TCI
Fara

Synonyms: (S)-(-)-2-Isocyanato-4-methylvaleric Acid Methyl Ester

I0467-25G I0467-5G
TCIAI0467-25GEA 20000 SEK
TCIAI0467-25G TCIAI0467-5G
Methyl (S)-(-)-2-isocyanato-4-methylvalerate ≥98.0%
Methyl (S)-(-)-2-isocyanato-4-methylvalerate
Formel: C₈H₁₃NO₃
MW: 171.20 g/mol
Kokpunkt: 74°C
MDL Number: MFCD00191532
CAS nummer: 39570-63-3
UN: 2206
ADR: 6.1 III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR