Methyl (S)-(-)-2-isocyanato-3-methylbutyrate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Methyl (S)-(-)-2-Isocyanatoisovalerate, (S)-(-)-2-Isocyanato-3-methylbutyric Acid Methyl Ester
I0466-1G
I0466-5G
TCIAI0466-1GEA
1025
SEK
TCIAI0466-1G
TCIAI0466-5G
Methyl (S)-(-)-2-isocyanato-3-methylbutyrate ≥98.0%
Methyl (S)-(-)-2-isocyanato-3-methylbutyrate
Formel:
C₇H₁₁NO₃ MW: 157,17 g/mol Kokpunkt: 76°C Flampunkt: 78°C |
MDL Number:
MFCD00191531 CAS nummer: 30293-86-8 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -18.0 to -23.0 deg (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...