4,5-Bis(hydroxymethyl)imidazole ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1H-Imidazole-4,5-dimethanol

I0457-1G I0457-5G
TCIAI0457-1GEA 1925 SEK
TCIAI0457-1G TCIAI0457-5G
4,5-Bis(hydroxymethyl)imidazole ≥98.0%
4,5-Bis(hydroxymethyl)imidazole
Formel: C₅H₈N₂O₂
MW: 128,13 g/mol
MDL Number: MFCD00191661
CAS nummer: 33457-48-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency
Melting point 159.0 to 165.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR