Diethyl isopropylidenesuccinate ≥98.0%

Leverantör: TCI

Synonyms: Isopropylidenesuccinic Acid Diethyl Ester

I0404-1G I0404-5G
TCIAI0404-1GEA 965 SEK
TCIAI0404-1G TCIAI0404-5G
Diethyl isopropylidenesuccinate ≥98.0%
Diethyl isopropylidenesuccinate
Formel: C₁₁H₁₈O₄
Kokpunkt: 103°C
MDL Number: MFCD00130132
CAS nummer: 42103-98-0

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR