N-Isopropylacrylamide ≥98.0% stabilised
Leverantör: TCI EUROPE
|
Varning
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
I0401-25G
I0401-500G
I0401-100G
TCIAI0401-25GEA
390
SEK
TCIAI0401-25G
TCIAI0401-500G
TCIAI0401-100G
N-Isopropylacrylamide ≥98.0% stabilised
N-Isopropylacrylamide
Formel:
C₆H₁₁NO MW: 113,16 g/mol Kokpunkt: 110°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00041913 CAS nummer: 2210-25-5 EINECS: 218-638-5 |
Specifikation testresultat
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 64.0 to 67.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...