2-Isopropenyltoluene ≥98.0% stabilised
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-Methyl-2-(prop-1-en-2-yl)benzene
Stabilization:
Stabilised med BHT/Ionol
I0298-1ML
I0298-5ML
TCIAI0298-1MLEA
2050
SEK
TCIAI0298-1ML
TCIAI0298-5ML
2-Isopropenyltoluene ≥98.0% stabilised
2-Isopropenyltoluene
Formel:
C₁₀H₁₂ MW: 132,21 g/mol Kokpunkt: 173°C Flampunkt: 43°C |
MDL Number:
MFCD00049092 CAS nummer: 7399-49-7 EINECS: 230-996-4 UN: 3295 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...