Isobutyraldehyde diethyl acetal ≥96.0%
Leverantör: TCI
|
Varning
|
Synonyms:
1,1-Diethoxy-2-methylpropane, 1,1-Diethoxyisobutane
I0287-25ML
I0287-5ML
TCIAI0287-25MLEA
1650
SEK
TCIAI0287-25ML
TCIAI0287-5ML
Isobutyraldehyde diethyl acetal ≥96.0%
Isobutyraldehyde diethyl acetal
Formel:
C₈H₁₈O₂ Kokpunkt: 138°C |
MDL Number:
MFCD00059384 CAS nummer: 1741-41-9 EINECS: 217-107-5 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...