2-Ethoxyethyl isobutyrate ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Isobutyric Acid 2-Ethoxyethyl Ester, Cellosolve Isobutyrate, Ethylene Glycol Monoethyl Ether Isobutyrate
I0282-25ML I0282-5ML
TCIAI0282-25MLEA 1400 SEK
TCIAI0282-25ML TCIAI0282-5ML
2-Ethoxyethyl isobutyrate ≥98.0%
2-Ethoxyethyl isobutyrate
Formel: C₈H₁₆O₃
MW: 160,21 g/mol
Kokpunkt: 85°C
Flampunkt: 57°C
MDL Number: MFCD00009250
CAS nummer: 54396-97-3
EINECS: 259-146-0
UN: 3272
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %