Monomethyl itaconate ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Itaconic Acid Monomethyl Ester, beta-Methyl Itaconate, 4-Methyl Methylenesuccinate, 4-Methoxy-2-methylene-4-oxobutanoic acid
I0269-25G
I0269-500G
I0269-100G
TCIAI0269-25GEA
655
SEK
TCIAI0269-25G
TCIAI0269-500G
TCIAI0269-100G
Monomethyl itaconate ≥98.0% (by GC, titration analysis)
Monomethyl itaconate
Formel:
C₆H₈O₄ MW: 144,13 g/mol Kokpunkt: 149°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00021713 CAS nummer: 7338-27-4 EINECS: 230-853-6 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 68.0 to 73.0 deg-C |
Solubility in Methanol | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...