Diethyl itaconate ≥98.0% stabilised
Leverantör: TCI EUROPE
Synonyms: Diethyl 2-methylenesuccinate, Itaconic Acid Diethyl Ester
Stabilization: med 4-tert-Butylpyrokatekol
I0204-500G I0204-25G
TCIAI0204-500GEA 6650 SEK
TCIAI0204-500G TCIAI0204-25G
Diethyl itaconate ≥98.0% stabilised
Diethyl itaconate
Formel: C₉H₁₄O₄
MW: 186,21 g/mol
Kokpunkt: 228°C
Flampunkt: 108°C
MDL Number: MFCD00059230
CAS nummer: 2409-52-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f11d15ef52d3853220afd1a9f411ce4ca6ff9520&vid=f11d15ef52d3853220afd1a9f411ce4ca6ff9520&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %