(E)-4-Hexen-3-one ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: (E)-Hex-4-en-3-one
H1735-25ML H1735-5ML
TCIAH1735-25MLEA 1075 SEK
TCIAH1735-25ML TCIAH1735-5ML
(E)-4-Hexen-3-one ≥98.0%
(E)-4-Hexen-3-one
Formel: C₆H₁₀O
MW: 98,15 g/mol
Kokpunkt: 137°C
MDL Number: MFCD00010631
CAS nummer: 50396-87-7
UN: 1224
ADR: 3,III
Order Now
Specifikation testresultat
Appearance White to Light yellow clear liquid
Purity (GC) min. 98.0 %