2-Hydroxy-5-n-octanoylbenzoic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 5-n-Octanoylsalicylic acid, 2-Hydroxy-5-octanoylbenzoic acid

H1712-5G H1712-25G
TCIAH1712-5GEA 510 SEK
TCIAH1712-5G TCIAH1712-25G
2-Hydroxy-5-n-octanoylbenzoic acid ≥98.0% (by GC, titration analysis)
2-Hydroxy-5-n-octanoylbenzoic acid
Formel: C₁₅H₂₀O₄
MW: 264.32 g/mol
MDL Number: MFCD11846247
CAS nummer: 78418-01-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 113.0 to 117.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR