7-Hydroxy-1-tetralone ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 7-Hydroxy-3,4-dihydronaphthalen-1(2H)-one

H1650-1G H1650-5G
TCIAH1650-1GEA 270 SEK
TCIAH1650-1G TCIAH1650-5G
7-Hydroxy-1-tetralone ≥98.0% (by GC, titration analysis)
7-Hydroxy-1-tetralone
Formel: C₁₀H₁₀O₂
MW: 162,19 g/mol
Kokpunkt: 215°C
MDL Number: MFCD01312225
CAS nummer: 22009-38-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63676deda6be2c754abcc1e2d824269c9c1db078&vid=63676deda6be2c754abcc1e2d824269c9c1db078&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 164.0 to 168.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR