Hederagenin ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 3beta,4alpha-3,23-Dihydroxyolean-12-en-28-oic acid

H1645-50MG H1645-200MG
TCIAH1645-50MGEA 1850 SEK
TCIAH1645-50MG TCIAH1645-200MG
Hederagenin ≥98.0% (by HPLC, titration analysis)
Hederagenin
Formel: C₃₀H₄₈O₄
MW: 472,71 g/mol
Smältpunkt: 334°C
Storage Temperature: Kylskåp
MDL Number: MFCD00017385
CAS nummer: 465-99-6
EINECS: 207-369-9
Merck Index: 14,04624
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f80af206e121697b53a7c6b2ca2de39df59890&vid=16f80af206e121697b53a7c6b2ca2de39df59890&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +81.0 to +85.0 deg (C=1, Pyridine)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR