4'-Hydroxyflavone ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-(4-Hydroxyphenyl)-4H-chromen-4-one, 2-(4-Hydroxyphenyl)-4H-1-benzopyran-4-one
H1638-1G
H1638-5G
TCIAH1638-1GEA
1325
SEK
TCIAH1638-1G
TCIAH1638-5G
4'-Hydroxyflavone ≥98.0% (by GC, titration analysis)
4'-Hydroxyflavone
Formel:
C₁₅H₁₀O₃ MW: 238,24 g/mol |
MDL Number:
MFCD00017687 CAS nummer: 4143-63-9 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...