1,1,1,5,5,5-Hexamethyl-3-[(trimethylsilyl)oxy]-3-vinyltrisiloxane ≥98.0%

Leverantör: TCI
Varning

Synonyms: 1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)-3-vinyltrisiloxane, Tris(trimethylsiloxy)(vinyl)silane

H1613-5ML H1613-25ML
TCIAH1613-5MLEA 685 SEK
TCIAH1613-5ML TCIAH1613-25ML
1,1,1,5,5,5-Hexamethyl-3-[(trimethylsilyl)oxy]-3-vinyltrisiloxane ≥98.0%
1,1,1,5,5,5-Hexamethyl-3-[(trimethylsilyl)oxy]-3-vinyltrisiloxane
Formel: C₁₁H₃₀O₃Si₄
MW: 322.70 g/mol
Kokpunkt: 88°C
Flampunkt: 66°C
MDL Number: MFCD00054159
CAS nummer: 5356-84-3
EINECS: 226-342-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e501f1b57f6935901f3c8f869e74b46b7777d58&vid=6e501f1b57f6935901f3c8f869e74b46b7777d58&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR