3-Hydroxycoumarin 98,0 GC, T
Leverantör: TCI
|
Varning
|
Synonyms:
3-Hydroxy-2-benzopyrone, 3-Hydroxy-2H-chromen-2-one
H1585-1G
H1585-5G
TCIAH1585-1GEA
1025
SEK
TCIAH1585-1G
TCIAH1585-5G
3-Hydroxycoumarin 98,0 GC, T
3-Hydroxycoumarin
Formel:
C₉H₆O₃ MW: 162.14 g/mol |
MDL Number:
MFCD00017490 CAS nummer: 939-19-5 EINECS: 213-355-3 |
|
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 151.0 to 155.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...