2'-Hydroxy-4',6'-dimethoxy-2-phenylacetophenone ≥94.0%
Leverantör: TCI EUROPE
Synonyms:
Benzyl 2-Hydroxy-4,6-dimethoxyphenyl Ketone
H1577-1G
H1577-5G
TCIAH1577-1GEA
1225
SEK
TCIAH1577-1G
TCIAH1577-5G
2'-Hydroxy-4',6'-dimethoxy-2-phenylacetophenone ≥94.0%
2'-Hydroxy-4',6'-dimethoxy-2-phenylacetophenone
Formel:
C₁₆H₁₆O₄ MW: 272,30 g/mol |
MDL Number:
MFCD00218596 CAS nummer: 39604-66-5 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 94.0 % |
Melting point | 114.0 to 119.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...