2-Hydroxy-4-methylbenzophenone ≥97.0%
Leverantör: TCI
|
Varning
|
H1575-5G
H1575-25G
TCIAH1575-5GEA
645
SEK
TCIAH1575-5G
TCIAH1575-25G
2-Hydroxy-4-methylbenzophenone ≥97.0%
2-Hydroxy-4-methylbenzophenone
Formel:
C₁₄H₁₂O₂ Kokpunkt: 234°C |
MDL Number:
MFCD01099035 CAS nummer: 3098-18-8 |
|
Specifikation testresultat
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...